Product Details

SNP ID
rs199571416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123805718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTATGTACACGTAGGACCCAGTA[A/G]TGAATGCCAGGGAGCTCAGGATGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6M1 PubMed Links

Gene Details

Gene
OR6M1
Gene Name
olfactory receptor family 6 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005325.1 632 Missense Mutation ACT,ATT T211I NP_001005325.1

View Full Product Details