Product Details

SNP ID
rs200795587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112179366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCATGTGTTGCACCGCTGCTGAC[C/T]ACAGTGGAAGAGGCTCCACGGGGCA
Phenotype
MIM: 611740 MIM: 605791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCO2 PubMed Links

Gene Details

Gene
BCO2
Gene Name
beta-carotene oxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037290.2 294 Silent Mutation ACC,ACT T25T NP_001032367.2
NM_001256397.1 294 Silent Mutation ACC,ACT T25T NP_001243326.1
NM_001256398.1 294 Silent Mutation ACC,ACT T59T NP_001243327.1
NM_001256400.1 294 Intron NP_001243329.1
NM_031938.5 294 Silent Mutation ACC,ACT T59T NP_114144.4
Gene
TEX12
Gene Name
testis expressed 12
There are no transcripts associated with this gene.

View Full Product Details