Product Details

SNP ID
rs200846527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18396946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTTGGTGCTGTTGGCATGGCCT[C/G]TGCCATCAGTATCTTAATGAAGGTA
Phenotype
MIM: 150000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LDHA PubMed Links

Gene Details

Gene
LDHA
Gene Name
lactate dehydrogenase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135239.1 473 Missense Mutation TCT,TGT S35C NP_001128711.1
NM_001165414.1 473 Missense Mutation TCT,TGT S64C NP_001158886.1
NM_001165415.1 473 Missense Mutation TCT,TGT S35C NP_001158887.1
NM_001165416.1 473 Missense Mutation TCT,TGT S35C NP_001158888.1
NM_005566.3 473 Missense Mutation TCT,TGT S35C NP_005557.1
Gene
MIR3159
Gene Name
microRNA 3159
There are no transcripts associated with this gene.

View Full Product Details