Product Details
- SNP ID
-
rs200916477
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:40114490 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTTGTGTGGTTGAAGGGAGATTT[A/G]TATGAGTTATAGTGATTTAGGTGCT
- Phenotype
-
MIM: 608817
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC4C
PubMed Links
Gene Details
- Gene
- LRRC4C
- Gene Name
- leucine rich repeat containing 4C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001258419.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
NP_001245348.1 |
NM_020929.2 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
NP_065980.1 |
XM_011520238.2 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_011518540.1 |
XM_011520239.2 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_011518541.1 |
XM_011520240.2 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_011518542.1 |
XM_011520241.2 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_011518543.1 |
XM_011520242.2 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_011518544.1 |
XM_011520243.2 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_011518545.1 |
XM_011520244.2 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_011518546.1 |
XM_017018070.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873559.1 |
XM_017018071.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873560.1 |
XM_017018072.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873561.1 |
XM_017018073.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873562.1 |
XM_017018074.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873563.1 |
XM_017018075.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873564.1 |
XM_017018076.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873565.1 |
XM_017018077.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873566.1 |
XM_017018078.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873567.1 |
XM_017018079.1 |
3027 |
Silent Mutation |
TAC,TAT |
Y601Y |
XP_016873568.1 |
View Full Product Details