Product Details

SNP ID
rs200916919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65377038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTGGCTCTGTCTGGTTTTGTAG[A/C]CGGACTTTGATGAGGTCAAAAGGAG
Phenotype
MIM: 610825
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FRMD8 PubMed Links

Gene Details

Gene
FRMD8
Gene Name
FERM domain containing 8
There are no transcripts associated with this gene.

Gene
SLC25A45
Gene Name
solute carrier family 25 member 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077241.2 1387 Silent Mutation NP_001070709.2
NM_001278250.2 1387 Silent Mutation NP_001265179.2
NM_001278251.2 1387 Silent Mutation NP_001265180.2
NM_001300820.1 1387 Silent Mutation NP_001287749.1
NM_182556.3 1387 Silent Mutation NP_872362.3
XM_006718509.3 1387 Silent Mutation XP_006718572.1
XM_006718510.3 1387 Silent Mutation XP_006718573.1
XM_011544943.2 1387 Silent Mutation XP_011543245.1
XM_011544944.2 1387 Silent Mutation XP_011543246.1
XM_011544947.2 1387 Silent Mutation XP_011543249.1
XM_011544949.2 1387 Silent Mutation XP_011543251.1
XM_017017562.1 1387 Silent Mutation XP_016873051.1
XM_017017563.1 1387 Silent Mutation XP_016873052.1
XM_017017564.1 1387 Silent Mutation XP_016873053.1
XM_017017565.1 1387 Silent Mutation XP_016873054.1
XM_017017566.1 1387 Silent Mutation XP_016873055.1
XM_017017567.1 1387 Silent Mutation XP_016873056.1
XM_017017568.1 1387 Silent Mutation XP_016873057.1
XM_017017569.1 1387 Silent Mutation XP_016873058.1
XM_017017570.1 1387 Silent Mutation XP_016873059.1
XM_017017571.1 1387 Silent Mutation XP_016873060.1

View Full Product Details