Product Details

SNP ID
rs200982548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58358295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGGAATGACTGGAACTTGGTC[A/G]TATGTACATGATGATGACAGTTATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5B17 PubMed Links

Gene Details

Gene
OR5B17
Gene Name
olfactory receptor family 5 subfamily B member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005489.1 775 Nonsense Mutation CGA,TGA R259* NP_001005489.1

View Full Product Details