Product Details

SNP ID
rs201029324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35432498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGGCTCAGGAGATGCTCGTCCC[A/G]GGAAGGACACAGCCGCGATGCCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAMR1 PubMed Links

Gene Details

Gene
PAMR1
Gene Name
peptidase domain containing associated with muscle regeneration 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001991.2 2518 Missense Mutation CCG,CTG P674L NP_001001991.1
NM_001282675.1 2518 Missense Mutation CCG,CTG P634L NP_001269604.1
NM_001282676.1 2518 Missense Mutation CCG,CTG P563L NP_001269605.1
NM_015430.3 2518 Missense Mutation CCG,CTG P691L NP_056245.2

View Full Product Details