Product Details

SNP ID
rs200984341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5422379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACAGAGCCATCTGTCCACCAGC[A/G]CATGTATCTGTTTCTCTCCATGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 269 Intron NP_001005567.2
Gene
OR51Q1
Gene Name
olfactory receptor family 51 subfamily Q member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004757.2 269 Missense Mutation CAC,CGC H60R NP_001004757.1

View Full Product Details