Product Details

SNP ID
rs200762365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108160953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGAAGGACGAGAGTTTCGCTCA[C/T]GTTCATCTAATTCCTCAATTTTCCG
Phenotype
MIM: 601448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPAT PubMed Links

Gene Details

Gene
NPAT
Gene Name
nuclear protein, coactivator of histone transcription
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321307.1 5000 Missense Mutation CAT,CGT H1385R NP_001308236.1
NM_002519.2 5000 Missense Mutation CAT,CGT H1378R NP_002510.2
XM_011542854.2 5000 Missense Mutation CAT,CGT H1387R XP_011541156.1
XM_017017802.1 5000 Missense Mutation CAT,CGT H1319R XP_016873291.1
XM_017017803.1 5000 Missense Mutation CAT,CGT H1319R XP_016873292.1

View Full Product Details