Product Details

SNP ID
rs201032623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116821041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGTCCTTCTCCAGGAAGCTCAA[C/G]TGGCCTTCCACGTCCCCCGCATGGG
Phenotype
MIM: 107690 MIM: 107720
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOA4 PubMed Links

Gene Details

Gene
APOA4
Gene Name
apolipoprotein A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000482.3 1121 Missense Mutation CAC,CAG H339Q NP_000473.2
Gene
APOC3
Gene Name
apolipoprotein C3
There are no transcripts associated with this gene.

View Full Product Details