Product Details

SNP ID
rs201011553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:104948552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACGTGGCGTTGAAGAGCAGAA[A/G]GCAATGAAGTCCTTCTCCACGTGGG
Phenotype
MIM: 602664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP4 PubMed Links

Gene Details

Gene
CASP4
Gene Name
caspase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001225.3 979 Silent Mutation GCC,GCT A302A NP_001216.1
NM_033306.2 979 Silent Mutation GCC,GCT A246A NP_150649.1
XM_011543019.1 979 Silent Mutation GCC,GCT A211A XP_011541321.1
XM_017018397.1 979 Silent Mutation GCC,GCT A211A XP_016873886.1

View Full Product Details