Product Details

SNP ID
rs201034217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95979245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGACTGATTCAAATTAGGCAA[A/C]ATCTGTGTGCTTAGCTGATTGGGTC
Phenotype
MIM: 607537
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAML2 PubMed Links

Gene Details

Gene
MAML2
Gene Name
mastermind like transcriptional coactivator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032427.3 6294 Missense Mutation ATG,ATT M1058I NP_115803.1
XM_011543023.2 6294 Missense Mutation ATG,ATT M911I XP_011541325.1
XM_011543024.2 6294 Missense Mutation ATG,ATT M830I XP_011541326.1
XM_011543025.1 6294 Intron XP_011541327.1

View Full Product Details