Product Details

SNP ID
rs201057349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75720169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATGAAGGACTATTTCCCCATCT[C/T]GGTGAGTATTGAGGCTGGTTGGGGT
Phenotype
MIM: 610270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MOGAT2 PubMed Links

Gene Details

Gene
MOGAT2
Gene Name
monoacylglycerol O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025098.3 339 Missense Mutation TCG,TTG S90L NP_079374.2
XM_011545267.1 339 Missense Mutation TCG,TTG S90L XP_011543569.1

View Full Product Details