Product Details

SNP ID
rs200311856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67393695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACCCTATCGCCCATCCAGGGGT[A/G]CGGAAGACTCACAACCTGTCCTTCC
Phenotype
MIM: 176875 MIM: 603761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100130987 PubMed Links

Gene Details

Gene
LOC100130987
Gene Name
uncharacterized LOC100130987
There are no transcripts associated with this gene.

Gene
PPP1CA
Gene Name
protein phosphatase 1 catalytic subunit alpha
There are no transcripts associated with this gene.

Gene
RAD9A
Gene Name
RAD9 checkpoint clamp component A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243224.1 659 Silent Mutation GTA,GTG V42V NP_001230153.1
NM_004584.2 659 Silent Mutation GTA,GTG V118V NP_004575.1
XM_006718652.3 659 Silent Mutation GTA,GTG V129V XP_006718715.1
XM_017018097.1 659 Intron XP_016873586.1
XM_017018098.1 659 UTR 5 XP_016873587.1

View Full Product Details