Product Details
- SNP ID
-
rs199973159
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:8911515 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCGACGTTCCCTGCAGCGTTCAGC[A/G]AGGCTGGCTCTAGAAGTGCTGGAGA
- Phenotype
-
MIM: 609191
MIM: 140750
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKIP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1133833] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKIP1
- Gene Name
- A-kinase interacting protein 1
- Gene
- C11orf16
- Gene Name
- chromosome 11 open reading frame 16
There are no transcripts associated with this gene.
- Gene
- ST5
- Gene Name
- suppression of tumorigenicity 5
View Full Product Details