Product Details

SNP ID
rs200017623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114297995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATTGACATCGGCTCTGGCCCC[A/G]CTATCTATCAGCTCCTCTCTGCTTG
Phenotype
MIM: 600008
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NNMT PubMed Links

Gene Details

Gene
NNMT
Gene Name
nicotinamide N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006169.2 943 Missense Mutation ACT,GCT T67A NP_006160.1

View Full Product Details