Product Details

SNP ID
rs200132677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59457339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTATGACAGATACCTTGCAATC[A/G]CCAAGCCCCTGCACTATGTGACCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D6 PubMed Links

Gene Details

Gene
OR4D6
Gene Name
olfactory receptor family 4 subfamily D member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004708.1 379 Missense Mutation ACC,GCC T127A NP_001004708.1

View Full Product Details