Product Details

SNP ID
rs200245947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124542777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGGGCAACTTTGACATCCTTG[C/T]TCCTCAAGCTATAGATTAATGGGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8B12 PubMed Links

Gene Details

Gene
OR8B12
Gene Name
olfactory receptor family 8 subfamily B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005195.1 878 Missense Mutation AAC,AGC N293S NP_001005195.1

View Full Product Details