Product Details

SNP ID
rs200133001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59124319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTGAAATGCAGAATCCAAATTT[C/G]AACAATAAAGAATGTTGTTTCACCT
Phenotype
MIM: 615584
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM111B PubMed Links

Gene Details

Gene
FAM111B
Gene Name
family with sequence similarity 111 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142703.1 465 Missense Mutation TTC,TTG F44L NP_001136175.1
NM_001142704.1 465 Missense Mutation TTC,TTG F44L NP_001136176.1
NM_198947.3 465 Missense Mutation TTC,TTG F74L NP_945185.1
Gene
LOC101927204
Gene Name
uncharacterized LOC101927204
There are no transcripts associated with this gene.

View Full Product Details