Product Details

SNP ID
rs200202420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70206322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAGAAGGAGAACGCAACAGTGG[C/T]CCACCTGGTGGGGGCTCTCAGGTCC
Phenotype
MIM: 602457
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FADD PubMed Links

Gene Details

Gene
FADD
Gene Name
Fas associated via death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003824.3 773 Missense Mutation GCC,GTC A159V NP_003815.1

View Full Product Details