Product Details
- SNP ID
-
rs200270292
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:48306543 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTTCTGCCCACTTCTTTGGTGG[C/T]ACTGAGATCTTCCTCCTTACAGCCA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR4S1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs202137071,rs571664984] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR4S1
- Gene Name
- olfactory receptor family 4 subfamily S member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004725.1 |
321 |
Silent Mutation |
GGC,GGT |
G107G |
NP_001004725.1 |
View Full Product Details