Product Details

SNP ID
rs200180340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59124617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAAAGAAGATGGACACATATTA[C/T]GCCAATGTGAAAATCCAAACATGGA
Phenotype
MIM: 615584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM111B PubMed Links

Gene Details

Gene
FAM111B
Gene Name
family with sequence similarity 111 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142703.1 763 Missense Mutation CGC,TGC R144C NP_001136175.1
NM_001142704.1 763 Missense Mutation CGC,TGC R144C NP_001136176.1
NM_198947.3 763 Missense Mutation CGC,TGC R174C NP_945185.1
Gene
LOC101927204
Gene Name
uncharacterized LOC101927204
There are no transcripts associated with this gene.

View Full Product Details