Product Details
- SNP ID
-
rs200180369
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:18025608 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTTTGAACAGCCTCCTCTGAAGC[A/G]CCAAGAGAAGCCAAGCCAATTTCTT
- Phenotype
-
MIM: 191060
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TPH1
PubMed Links
Gene Details
- Gene
- TPH1
- Gene Name
- tryptophan hydroxylase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004179.2 |
923 |
Silent Mutation |
GGC,GGT |
G299G |
NP_004170.1 |
View Full Product Details