Product Details

SNP ID
rs200181657
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124030510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGAGGAACAAGGAGGTGAAGAA[A/G]GCTCTGTTGAAGCTGAAAGACAAAG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR10G7 PubMed Links
Additional Information
For this assay, SNP(s) [rs11825515] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR10G7
Gene Name
olfactory receptor family 10 subfamily G member 7
There are no transcripts associated with this gene.

Gene
OR10G8
Gene Name
olfactory receptor family 10 subfamily G member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004464.1 Intron NP_001004464.1
Gene
OR10G9
Gene Name
olfactory receptor family 10 subfamily G member 9
There are no transcripts associated with this gene.

View Full Product Details