Product Details

SNP ID
rs200277918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2402690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCGTGGAGGGCGAACACGGGAC[A/G]GAGTATGACACGCTGCCTTCCGACA
Phenotype
MIM: 186845 MIM: 604600 MIM: 603852
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD81 PubMed Links

Gene Details

Gene
CD81
Gene Name
CD81 molecule
There are no transcripts associated with this gene.

Gene
TRPM5
Gene Name
transient receptor potential cation channel subfamily M member 5
There are no transcripts associated with this gene.

Gene
TSSC4
Gene Name
tumor suppressing subtransferable candidate 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297658.1 689 Silent Mutation ACA,ACG T19T NP_001284587.1
NM_001297659.1 689 Silent Mutation ACA,ACG T19T NP_001284588.1
NM_001297660.1 689 Silent Mutation ACA,ACG T19T NP_001284589.1
NM_001297661.1 689 Intron NP_001284590.1
NM_005706.3 689 Silent Mutation ACA,ACG T19T NP_005697.2
XM_006718118.2 689 Silent Mutation ACA,ACG T19T XP_006718181.1
XM_011519830.2 689 Silent Mutation ACA,ACG T19T XP_011518132.1

View Full Product Details