Product Details
- SNP ID
-
rs200192032
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:65850030 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATGAGGAGTACGCCGGCCTCAT[C/T]GTGAGGAGGGGCTGGGCAGGGGCTG
- Phenotype
-
MIM: 601442
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CFL1
PubMed Links
Gene Details
- Gene
- CFL1
- Gene Name
- cofilin 1
There are no transcripts associated with this gene.
- Gene
- SNX32
- Gene Name
- sorting nexin 32
View Full Product Details