Product Details

SNP ID
rs200214902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5581610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATCCACATCCTCCAAGCAGTCT[A/T]CCGCCTCCTTTCTCAAGATGCCCGC
Phenotype
MIM: 608487
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR52B6 PubMed Links

Gene Details

Gene
OR52B6
Gene Name
olfactory receptor family 52 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005162.2 734 Missense Mutation TAC,TTC Y245F NP_001005162.2
Gene
TRIM5
Gene Name
tripartite motif containing 5
There are no transcripts associated with this gene.

View Full Product Details