Product Details

SNP ID
rs200677353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17736116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCACGCGGCTCGCCTGGCTGGC[A/G]GAGCCCGACGCCCACAGCCACTTCG
Phenotype
MIM: 176258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNC1 PubMed Links

Gene Details

Gene
KCNC1
Gene Name
potassium voltage-gated channel subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112741.1 169 Silent Mutation GCA,GCG A38A NP_001106212.1
NM_004976.4 169 Silent Mutation GCA,GCG A38A NP_004967.1
XM_011520080.2 169 Silent Mutation GCA,GCG A38A XP_011518382.1

View Full Product Details