Product Details
- SNP ID
-
rs200402268
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:17393012 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTCACTTGTCTGCACGGACGAA[A/G]GAGGCGAAGACGCTGTCCTTCCGGC
- Phenotype
-
MIM: 600509
MIM: 600937
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCC8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8192690] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCC8
- Gene Name
- ATP binding cassette subfamily C member 8
- Gene
- KCNJ11
- Gene Name
- potassium voltage-gated channel subfamily J member 11
There are no transcripts associated with this gene.
View Full Product Details