Product Details

SNP ID
rs200426997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67436119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGGCTGGCCCAGAGCAGGCGCC[A/G]CGTGCGGCCCCACAGCGCGGCACCT
Phenotype
MIM: 609849 MIM: 601577 MIM: 608939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CORO1B PubMed Links

Gene Details

Gene
CORO1B
Gene Name
coronin 1B
There are no transcripts associated with this gene.

Gene
PTPRCAP
Gene Name
protein tyrosine phosphatase, receptor type C associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005608.2 298 Missense Mutation NP_005599.1
Gene
RPS6KB2
Gene Name
ribosomal protein S6 kinase B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003952.2 298 Intron NP_003943.2
XM_005274164.1 298 Intron XP_005274221.1
XM_005274165.4 298 Intron XP_005274222.1
XM_006718655.3 298 Intron XP_006718718.1
XM_006718656.3 298 Intron XP_006718719.1
XM_006718657.1 298 Intron XP_006718720.1
XM_017018108.1 298 Intron XP_016873597.1

View Full Product Details