Product Details

SNP ID
rs200475918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102692948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATCCATGCTTACCTTTGTACTG[A/G]AAAGCAGCATCAACACGGATACTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP27 PubMed Links

Gene Details

Gene
MMP27
Gene Name
matrix metallopeptidase 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022122.2 1310 Silent Mutation TTC,TTT F429F NP_071405.2
XM_011542948.2 1310 Silent Mutation TTC,TTT F386F XP_011541250.1
XM_011542949.2 1310 Silent Mutation TTC,TTT F375F XP_011541251.1
XM_011542950.2 1310 Silent Mutation TTC,TTT F324F XP_011541252.1
XM_017018120.1 1310 Silent Mutation TTC,TTT F428F XP_016873609.1

View Full Product Details