Product Details

SNP ID
rs200659555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43680844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAAGCCATGGAGAGCGCTCTCC[C/T]CGCCGCCGGCTTCCTGTACTGGGTC
Phenotype
MIM: 609574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD17B12 PubMed Links

Gene Details

Gene
HSD17B12
Gene Name
hydroxysteroid 17-beta dehydrogenase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016142.2 50 Missense Mutation CCC,CTC P6L NP_057226.1
XM_011520156.1 50 UTR 5 XP_011518458.1
XM_017017881.1 50 Intron XP_016873370.1

View Full Product Details