Product Details

SNP ID
rs200546421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4848271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACCCACCTTCCTGGGCTGCAAC[C/T]TGTTGAGTATTCTCTTTCTAATCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51S1 PubMed Links

Gene Details

Gene
OR51S1
Gene Name
olfactory receptor family 51 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004758.1 938 Missense Mutation AAG,AGG K313R NP_001004758.1

View Full Product Details