Product Details

SNP ID
rs200527660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118339851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGAGCAGAGTGGCAATGACATC[A/G]GTGACAATGATGCCAGCCACGGTGG
Phenotype
MIM: 186790 MIM: 186740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD3D PubMed Links

Gene Details

Gene
CD3D
Gene Name
CD3d molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000732.4 467 Silent Mutation ACC,ACT T110T NP_000723.1
NM_001040651.1 467 Intron NP_001035741.1
XM_017018543.1 467 Silent Mutation ACC,ACT T110T XP_016874032.1
Gene
CD3G
Gene Name
CD3g molecule
There are no transcripts associated with this gene.

View Full Product Details