Product Details
- SNP ID
-
rs200341456
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:56612619 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTTGCATGGCAAGGATTACATCT[C/G]TGTTCCGTAGGCTATAGATCAATGG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR5M1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4939078] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR5M1
- Gene Name
- olfactory receptor family 5 subfamily M member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004740.1 |
884 |
Missense Mutation |
ACA,AGA |
T295R |
NP_001004740.1 |
View Full Product Details