Product Details

SNP ID
rs200575431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:120330232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTCTCCGGGAAACAGGGCACT[A/G]TCACAGTTTCACTGGAGATGTAGTG
Phenotype
MIM: 604763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF12 PubMed Links

Gene Details

Gene
ARHGEF12
Gene Name
Rho guanine nucleotide exchange factor 12
There are no transcripts associated with this gene.

Gene
TMEM136
Gene Name
transmembrane protein 136
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198670.1 685 Missense Mutation TAT,TGT Y174C NP_001185599.1
NM_001198671.1 685 Missense Mutation TAT,TGT Y152C NP_001185600.1
NM_001198672.1 685 Missense Mutation TAT,TGT Y51C NP_001185601.1
NM_001198673.1 685 Missense Mutation TAT,TGT Y51C NP_001185602.1
NM_001198674.1 685 Intron NP_001185603.1
NM_001198675.1 685 Missense Mutation TAT,TGT Y51C NP_001185604.1
NM_174926.2 685 Intron NP_777586.1
XM_017017327.1 685 Intron XP_016872816.1

View Full Product Details