Product Details

SNP ID
rs200487186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118527595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGACCTCGGAGAGGAAGCAGAAA[A/C]GGAAGAACGAGAAGAAGGTGGGCAT
Phenotype
MIM: 159555 MIM: 613934
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KMT2A PubMed Links

Gene Details

Gene
KMT2A
Gene Name
lysine methyltransferase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197104.1 101 Intron NP_001184033.1
NM_005933.3 101 Intron NP_005924.2
XM_006718839.3 101 Intron XP_006718902.2
XM_011542829.2 101 Intron XP_011541131.1
XM_011542830.2 101 Intron XP_011541132.1
XM_011542831.2 101 Intron XP_011541133.1
XM_011542833.2 101 Intron XP_011541135.1
Gene
LOC101929089
Gene Name
uncharacterized LOC101929089
There are no transcripts associated with this gene.

Gene
TMEM25
Gene Name
transmembrane protein 25
There are no transcripts associated with this gene.

Gene
TTC36
Gene Name
tetratricopeptide repeat domain 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080441.1 101 Missense Mutation AAG,ACG K34T NP_001073910.1

View Full Product Details