Product Details

SNP ID
rs200466437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:799377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCGGGGTCCTTGGGGGCCAAG[C/T]CCATGCGTCGGATGCTGTCCTGGTA
Phenotype
MIM: 608213 MIM: 605247 MIM: 609302
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEND1 PubMed Links

Gene Details

Gene
CEND1
Gene Name
cell cycle exit and neuronal differentiation 1
There are no transcripts associated with this gene.

Gene
PANO1
Gene Name
proapoptotic nucleolar protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293167.1 3820 Intron NP_001280096.1
Gene
PIDD1
Gene Name
p53-induced death domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145886.3 3820 Missense Mutation GAC,GGC D888G NP_665893.2
NM_145887.3 3820 Missense Mutation GAC,GGC D871G NP_665894.2
XM_005253005.4 3820 Missense Mutation GAC,GGC D912G XP_005253062.1
XM_005253006.4 3820 Missense Mutation GAC,GGC D912G XP_005253063.1
XM_005253007.3 3820 Missense Mutation GAC,GGC D912G XP_005253064.1
XM_005253008.4 3820 Missense Mutation GAC,GGC D766G XP_005253065.1
XM_011520209.2 3820 Missense Mutation GAC,GGC D898G XP_011518511.1
XM_011520210.2 3820 Missense Mutation GAC,GGC D888G XP_011518512.1
XM_011520211.2 3820 Missense Mutation GAC,GGC D871G XP_011518513.1
XM_011520212.1 3820 Missense Mutation GAC,GGC D766G XP_011518514.1
XM_011520213.2 3820 Missense Mutation GAC,GGC D599G XP_011518515.1
XM_017017993.1 3820 Missense Mutation GAC,GGC D575G XP_016873482.1
Gene
SLC25A22
Gene Name
solute carrier family 25 member 22
There are no transcripts associated with this gene.

View Full Product Details