Product Details

SNP ID
rs200490684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56664049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTGGCAGCCATGGCCTATGGT[C/T]GTTTTGTGGCCATTTGTCGACCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5AR1 PubMed Links

Gene Details

Gene
OR5AR1
Gene Name
olfactory receptor family 5 subfamily AR member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004730.1 364 Missense Mutation CGT,TGT R122C NP_001004730.1

View Full Product Details