Product Details

SNP ID
rs200468825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17719847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCCCCCGACGGCTCTCTCTGCT[C/T]CTTTGCCACAACGGACGACTTCTAT
Phenotype
MIM: 159970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYOD1 PubMed Links

Gene Details

Gene
MYOD1
Gene Name
myogenic differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002478.4 285 Missense Mutation TCC,TTC S22F NP_002469.2

View Full Product Details