Product Details

SNP ID
rs200561246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58228667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTAGCAGAGTTCCAGGAAAGAC[A/G]GGTTGGACAGGAAGAAATACATCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10Q1 PubMed Links

Gene Details

Gene
OR10Q1
Gene Name
olfactory receptor family 10 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004471.2 252 Missense Mutation CCG,CTG P70L NP_001004471.1

View Full Product Details