Product Details

SNP ID
rs199669171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67056929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGGCGGCCCAGGCCGCGGGTGA[A/G]GAGGCGCCACCAGGCGTGCGGTCCG
Phenotype
MIM: 605781 MIM: 606436
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHOD PubMed Links

Gene Details

Gene
RHOD
Gene Name
ras homolog family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300886.1 112 Silent Mutation GAA,GAG E9E NP_001287815.1
NM_014578.3 112 Silent Mutation GAA,GAG E9E NP_055393.1
Gene
SYT12
Gene Name
synaptotagmin 12
There are no transcripts associated with this gene.

View Full Product Details