Product Details

SNP ID
rs199718040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59364687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCATAGATGTCTGCTATATCAGC[C/T]CCACAGTCCCCAAGATGCTCTCCAA
Phenotype
MIM: 615702
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5AN1 PubMed Links

Gene Details

Gene
OR5AN1
Gene Name
olfactory receptor family 5 subfamily AN member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004729.1 229 Missense Mutation CCC,TCC P77S NP_001004729.1

View Full Product Details