Product Details

SNP ID
rs199948760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:373028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAACAGTCACTGCCCCCCCAGAC[A/G]CGCACCACCGTGAAGAAGTTGGCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALNT4 PubMed Links

Gene Details

Gene
B4GALNT4
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178537.4 456 Silent Mutation ACA,ACG T149T NP_848632.2
XM_017017654.1 456 Silent Mutation ACA,ACG T57T XP_016873143.1

View Full Product Details