Product Details

SNP ID
rs199744312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126271571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGTGATCGTGGGAGGTGGGGTG[C/T]TTGGCTTGTCTGTGGCCTATTGGCT
Phenotype
MIM: 616587 MIM: 613622 MIM: 182180 MIM: 606252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
There are no transcripts associated with this gene.

Gene
FOXRED1
Gene Name
FAD dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017547.3 1519 Missense Mutation CTT,TTT L74F NP_060017.1
XM_006718879.3 1519 Intron XP_006718942.1
XM_006718881.3 1519 Intron XP_006718944.1
XM_011542895.2 1519 UTR 5 XP_011541197.1
XM_011542896.2 1519 UTR 5 XP_011541198.1
XM_017018000.1 1519 Missense Mutation CTT,TTT L74F XP_016873489.1
XM_017018001.1 1519 UTR 5 XP_016873490.1
XM_017018002.1 1519 Intron XP_016873491.1
XM_017018003.1 1519 UTR 5 XP_016873492.1
XM_017018004.1 1519 UTR 5 XP_016873493.1
XM_017018005.1 1519 UTR 5 XP_016873494.1
XM_017018006.1 1519 UTR 5 XP_016873495.1
Gene
SRPRA
Gene Name
SRP receptor alpha subunit
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
There are no transcripts associated with this gene.

View Full Product Details