Product Details

SNP ID
rs199698252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56318667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATTCTCTCAGCCATGTCCTAC[A/G]ACCTCTATGTGGCCATCTGTAACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8K3 PubMed Links

Gene Details

Gene
OR8K3
Gene Name
olfactory receptor family 8 subfamily K member 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005202.1 361 Missense Mutation AAC,GAC N121D NP_001005202.1

View Full Product Details