Product Details
- SNP ID
-
rs199698372
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:128462496 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTCTTGTGGATGATGTTTTTGTC[A/G]TAATAGTAGCGTAGGCCACGGCTCA
- Phenotype
-
MIM: 164720
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ETS1
PubMed Links
Gene Details
- Gene
- ETS1
- Gene Name
- ETS proto-oncogene 1, transcription factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143820.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y441Y |
NP_001137292.1 |
NM_001162422.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y181Y |
NP_001155894.1 |
NM_005238.3 |
1182 |
Silent Mutation |
TAC,TAT |
Y397Y |
NP_005229.1 |
XM_011542649.2 |
1182 |
Silent Mutation |
TAC,TAT |
Y475Y |
XP_011540951.1 |
XM_011542650.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y431Y |
XP_011540952.1 |
XM_011542651.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y388Y |
XP_011540953.1 |
XM_011542652.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y310Y |
XP_011540954.1 |
XM_017017314.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y441Y |
XP_016872803.1 |
XM_017017315.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y406Y |
XP_016872804.1 |
XM_017017316.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y374Y |
XP_016872805.1 |
XM_017017317.1 |
1182 |
Silent Mutation |
TAC,TAT |
Y354Y |
XP_016872806.1 |
View Full Product Details