Product Details

SNP ID
rs199700131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26995111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGCTGCTGGCCCTCACCATTAG[A/G]AGCCATGGGACCCGACTAGGTCGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FIBIN PubMed Links

Gene Details

Gene
FIBIN
Gene Name
fin bud initiation factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203371.1 1031 Silent Mutation AGA,AGG R195R NP_976249.1

View Full Product Details