Product Details

SNP ID
rs199792888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66284726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGGTAGAGCTGGAGACGCTGCC[A/G]GGGGACGGGGCCCCCCATGCTGTCG
Phenotype
MIM: 611288 MIM: 612565 MIM: 611484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNIH2 PubMed Links

Gene Details

Gene
CNIH2
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182553.2 1040 Intron NP_872359.1
Gene
RAB1B
Gene Name
RAB1B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM151A
Gene Name
transmembrane protein 151A
There are no transcripts associated with this gene.

Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300861.1 1040 Missense Mutation CGG,TGG R213W NP_001287790.1
NM_020470.2 1040 Missense Mutation CGG,TGG R265W NP_065203.2
XM_005273720.3 1040 Silent Mutation CCC,CCT P294P XP_005273777.1
XM_017017139.1 1040 Silent Mutation CCC,CCT P306P XP_016872628.1
XM_017017140.1 1040 Missense Mutation CGG,TGG R277W XP_016872629.1
XM_017017141.1 1040 Intron XP_016872630.1

View Full Product Details