Product Details

SNP ID
rs199655061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59831762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGCGATATTGTTGATAGAAGAGA[C/G]GACAAGGCCCCAAGATGTCATTGTG
Phenotype
MIM: 609342
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005142.2 1142 Missense Mutation CTC,GTC L370V NP_005133.2
XM_011544939.2 1142 Missense Mutation CTC,GTC L356V XP_011543241.1

View Full Product Details